siRNA Details
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317800.1 | NP_001304729.1 | 3 | 435 | ||
NM_003974.3 | NP_003965.2 | 5 | 805 | |||
NM_201349.2 | NP_958728.1 | 4 | 717 | |||
XM_005273680.4 | XP_005273737.1 | 4 | 532 | |||
GenBank mRNA | AF034970.1 | - | 5 | 698 | ||
AK226110.1 | - | 3 | 405 | |||
AK292126.1 | - | 5 | 761 | |||
AK296671.1 | - | 4 | 699 | |||
BC032623.1 | - | 5 | 773 | 5242017 | ||
CB270269.1 | - | 3 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
