siRNA Details
Gene Symbol: GDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000514.3 | NP_000505.1 | 3 | 787 | ||
NM_001190468.1 | NP_001177397.1 | 3 | 807 | |||
NM_001190469.1 | NP_001177398.1 | 3 | 729 | |||
NM_001278098.1 | NP_001265027.1 | 3 | 615 | |||
NM_199231.2 | NP_954701.1 | 3 | 632 | |||
XM_011514030.2 | XP_011512332.1 | 2 | 500 | |||
XM_017009337.1 | XP_016864826.1 | 3 | 1073 | |||
GenBank mRNA | AY052832.1 | - | 2 | 509 | ||
BC069119.1 | - | 2 | 587 | 7216973 | ||
BC069369.1 | - | 2 | 587 | 7262145 | ||
BC128108.1 | - | 2 | 587 | 40112122 | ||
BC128109.1 | - | 2 | 587 | 40112125 | ||
CR541923.1 | - | 2 | 587 | |||
DQ235474.1 | - | 3 | 589 | |||
KU178093.1 | - | 2 | 587 | |||
KU178094.1 | - | 2 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
