siRNA Details
Gene Symbol: RBM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291780.1 | NP_001278709.1 | 3 | 602 | ||
NM_017495.5 | NP_059965.2 | 2 | 506 | |||
NM_183425.2 | NP_906270.1 | 2 | 506 | |||
XM_005260446.3 | XP_005260503.1 | 3 | 540 | |||
XM_011528885.2 | XP_011527187.1 | 3 | 534 | |||
GenBank mRNA | AF432218.1 | - | 2 | 502 | ||
BC018711.1 | - | 2 | 258 | 4852523 | ||
BM549757.1 | - | 501 | ||||
DN998561.1 | - | 2 | 491 | |||
X75314.1 | - | 2 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
