siRNA Details
Gene Symbol: RBM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017495.5 | NP_059965.2 | 1, 2 | 423 | ||
NM_183425.2 | NP_906270.1 | 1, 2 | 423 | |||
GenBank mRNA | AF432218.1 | - | 1, 2 | 419 | ||
BC018711.1 | - | 1, 2 | 175 | 4852523 | ||
BM549757.1 | - | 418 | ||||
DB504051.1 | - | 1, 2 | 424 | |||
DN998561.1 | - | 1, 2 | 408 | |||
X75314.1 | - | 1, 2 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
