siRNA Details
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033035.4 | NP_149024.1 | 1 | 277 | ||
NR_045089.1 | 2 | 1482 | ||||
GenBank mRNA | AF338732.1 | - | 1 | 111 | ||
AY037115.1 | - | 1 | 277 | |||
BC040592.1 | - | 2 | 1483 | 5269764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
