siRNA Details
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033035.4 | NP_149024.1 | 4 | 890 | ||
NM_138551.4 | NP_612561.2 | 2 | 672 | |||
NR_045089.1 | 5 | 2095 | ||||
XM_011543698.1 | XP_011542000.1 | 3 | 494 | |||
GenBank mRNA | AL833421.1 | - | 2 | 652 | ||
BC016720.1 | - | 2 | 605 | |||
BC022894.1 | - | 2 | 644 | |||
BC040592.1 | - | 5 | 2096 | 5269764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
