siRNA Details
Gene Symbol: CLSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009566.2 | NP_001009566.1 | 8 | 1884 | ||
NM_001302883.1 | NP_001289812.1 | 8 | 1884 | |||
NM_014944.4 | NP_055759.3 | 7 | 1854 | |||
GenBank mRNA | AB020718.1 | - | 8 | 1884 | ||
AF438482.1 | - | 7 | 1820 | |||
AK000605.1 | - | 4 | 551 | |||
AK289798.1 | - | 7 | 1062 | |||
AK304813.1 | - | 8 | 1126 | |||
AY753301.1 | - | 7 | 1061 | |||
BC033902.2 | - | 7 | 1312 | 5263192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
