siRNA Details
Gene Symbol: LOC101060341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271793.4 | XP_005271850.1 | 3 | 864 | ||
XM_017012925.1 | XP_016868414.1 | 4 | 1068 | |||
XM_017012926.1 | XP_016868415.1 | 2 | 873 | |||
XM_017012927.1 | XP_016868416.1 | 3 | 885 |
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024497.2 | 8 | 1619 | |||
GenBank mRNA | AK128746.1 | - | 1 | 176 |
Gene Symbol: LINC01000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024368.1 | 5 | 1146 | |||
GenBank mRNA | AK302487.1 | - | 5 | 1167 | ||
BC062355.1 | - | 5 | 1065 | |||
BC131589.1 | - | 5 | 1065 |
Gene Symbol: LINC01001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK293878.1 | - | 3 | 963 |
Gene Symbol: LINC01061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037596.1 | 3 | 909 | |||
GenBank mRNA | AK294946.1 | - | 1 | 29 |
Gene Symbol: LINC01347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029401.1 | 18 | 3585 | |||
GenBank mRNA | AL137733.1 | - | 18 | 3585 |
Gene Symbol: LOC100132062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028325.1 | 3 | 935 |
Gene Symbol: LOC100132287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028322.1 | 3 | 935 | |||
GenBank mRNA | AK311126.1 | - | 1 | 109 |
Gene Symbol: LOC100133331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028327.1 | 3 | 935 |
Gene Symbol: LOC101929819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK302511.1 | - | 7 | 1415 |
Gene Symbol: LOC107986800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012928.1 | XP_016868417.1 | 3 | 830 | ||
XM_017012929.1 | XP_016868418.1 | 2 | 818 |
Gene Symbol: LOC729218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103825.1 | 3 | 899 | |||
NR_109983.1 | 10 | 1842 | ||||
GenBank mRNA | AK024248.1 | - | 10 | 1813 |
Gene Symbol: LOC729737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039983.2 | 3 | 1258 | |||
GenBank mRNA | AK294489.1 | - | 1 | 591 | ||
AK302573.1 | - | 1 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
