siRNA Details
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007468.2 | NP_001007469.1 | 6 | 892 | ||
NM_001317946.1 | NP_001304875.1 | 6 | 946 | |||
NM_003073.4 | NP_003064.2 | 6 | 919 | |||
XM_011530345.2 | XP_011528647.1 | 6 | 942 | |||
GenBank mRNA | AB017523.1 | - | 6 | 653 | ||
AJ011737.1 | - | 6 | 827 | |||
AJ011738.1 | - | 6 | 800 | |||
AK021419.1 | - | 6 | 825 | |||
AK024025.1 | - | 6 | 914 | |||
BC117114.1 | - | 6 | 708 | 40125665 | ||
BC143667.1 | - | 6 | 708 | 9052181 | ||
CB161660.1 | - | 4 | 392 | |||
CR456581.1 | - | 6 | 733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
