siRNA Details
Gene Symbol: AGRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305275.1 | NP_001292204.1 | 14 | 2553 | ||
NM_198576.3 | NP_940978.2 | 14 | 2553 | |||
XM_005244749.3 | XP_005244806.1 | 14 | 2574 | |||
XM_011541429.2 | XP_011539731.1 | 14 | 2574 | |||
GenBank mRNA | AB191264.1 | - | 14 | 2553 | ||
AF016903.1 | - | 14 | 2446 | |||
AK021586.1 | - | 8 | 1147 | |||
BC084578.1 | - | 7 | 951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
