siRNA Details
Gene Symbol: HSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002156.4 | NP_002147.2 | 3 | 404 | ||
NM_199440.1 | NP_955472.1 | 3 | 384 | |||
GenBank mRNA | AK298844.1 | - | 3 | 323 | ||
BC002676.2 | - | 3 | 314 | 3609039 | ||
KU178139.1 | - | 2 | 267 | |||
KU178141.1 | - | 2 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
