siRNA Details
Gene Symbol: HSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002156.4 | NP_002147.2 | 12 | 1960 | ||
NM_199440.1 | NP_955472.1 | 12 | 1940 | |||
GenBank mRNA | AK298844.1 | - | 11 | 1711 | ||
BC002676.2 | - | 12 | 1870 | 3609039 | ||
BC003030.1 | - | 12 | 1869 | 2821157 | ||
BC047350.1 | - | 11 | 2030 | |||
BC067082.1 | - | 12 | 1868 | 4025725 | ||
BC073746.1 | - | 12 | 1868 | 6372068 | ||
BX647145.1 | - | 11 | 2625 | |||
M22382.1 | - | 12 | 1868 | |||
M34664.1 | - | 12 | 1847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
