siRNA Details
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190233.1 | NP_001177162.1 | 3 | 272 | ||
NM_145261.3 | NP_660304.1 | 3 | 260 | |||
NR_033721.1 | 2 | 208 | ||||
NR_033722.1 | 3 | 260 | ||||
NR_033723.1 | 3 | 260 | ||||
GenBank mRNA | AI268262.1 | - | 3 | 259 | ||
AK307097.1 | - | 3 | 249 | |||
AK309019.1 | - | 2 | 161 | |||
AK311765.1 | - | 3 | 188 | |||
BC009702.1 | - | 3 | 212 | 3892297 | ||
BC073989.1 | - | 3 | 205 | 5760865 | ||
BE773017.1 | - | 3 | 227 | |||
BF666599.1 | - | 3 | 222 | |||
BG492744.1 | - | 2 | 144 | |||
BM701756.1 | - | 3 | 213 | |||
BM702203.1 | - | 3 | 213 | |||
CB052107.1 | - | 3 | 198 | |||
CB159371.1 | - | 3 | 272 | |||
DA158066.1 | - | 2 | 341 | |||
KU178861.1 | - | 2 | 88 | |||
KU178862.1 | - | 2 | 88 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
