siRNA Details
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190233.1 | NP_001177162.1 | 2 | 200 | ||
NM_145261.3 | NP_660304.1 | 2 | 188 | |||
NR_033722.1 | 2 | 188 | ||||
NR_033723.1 | 2 | 188 | ||||
GenBank mRNA | AI268262.1 | - | 2 | 187 | ||
AK307097.1 | - | 2 | 177 | |||
AK311765.1 | - | 2 | 116 | |||
BC009702.1 | - | 2 | 140 | 3892297 | ||
BC073989.1 | - | 2 | 133 | 5760865 | ||
BE773017.1 | - | 2 | 155 | |||
BF666599.1 | - | 2 | 150 | |||
BM701756.1 | - | 2 | 141 | |||
BM702203.1 | - | 2 | 141 | |||
CB052107.1 | - | 2 | 126 | |||
CB159371.1 | - | 2 | 200 | |||
DA158066.1 | - | 2 | 188 | |||
KU178861.1 | - | 1 | 16 | |||
KU178862.1 | - | 1 | 16 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
