siRNA Details
Gene Symbol: COL12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004370.5 | NP_004361.3 | 51 | 8061 | ||
NM_080645.2 | NP_542376.2 | 36 | 4569 | |||
XM_011535434.1 | XP_011533736.1 | 51 | 8205 | |||
XM_011535435.1 | XP_011533737.1 | 50 | 7932 | |||
XM_011535436.2 | XP_011533738.1 | 36 | 4924 | |||
XM_017010252.1 | XP_016865741.1 | 50 | 8457 | |||
GenBank mRNA | BC146868.1 | - | 36 | 4513 | 9056796 | |
U73778.1 | - | 51 | 8008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
