siRNA Details
Gene Symbol: COL12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004370.5 | NP_004361.3 | 50 | 7986 | ||
NM_080645.2 | NP_542376.2 | 35 | 4494 | |||
XM_011535434.1 | XP_011533736.1 | 50 | 8130 | |||
XM_011535435.1 | XP_011533737.1 | 49 | 7857 | |||
XM_011535436.2 | XP_011533738.1 | 35 | 4849 | |||
XM_017010252.1 | XP_016865741.1 | 49 | 8382 | |||
GenBank mRNA | BC146868.1 | - | 35 | 4438 | 9056796 | |
U73778.1 | - | 50 | 7933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
