siRNA Details
Gene Symbol: NARF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038618.2 | NP_001033707.1 | 7, 8 | 1059 | ||
NM_001083608.1 | NP_001077077.1 | 6, 7 | 757 | |||
NM_012336.3 | NP_036468.1 | 7, 8 | 901 | |||
XM_005256340.3 | XP_005256397.1 | 7, 8 | 1067 | |||
XM_006722278.3 | XP_006722341.1 | 6, 7 | 854 | |||
XM_011523541.2 | XP_011521843.1 | 7, 8 | 991 | |||
XM_011523543.2 | XP_011521845.1 | 8, 9 | 904 | |||
XM_017024449.1 | XP_016879938.1 | 8, 9 | 1651 | |||
XM_017024450.1 | XP_016879939.1 | 8, 9 | 1141 | |||
GenBank mRNA | AF128406.1 | - | 7, 8 | 852 | ||
AK000929.1 | - | 7, 8 | 854 | |||
AK056966.1 | - | 7, 8 | 1034 | |||
AK093717.1 | - | 1, 2 | 735 | |||
AK303155.1 | - | 7, 8 | 816 | |||
AL137729.1 | - | 4, 5 | 492 | |||
BC016440.1 | - | 6, 7 | 644 | 3447710 | ||
EF601656.1 | - | 1, 2 | 59 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
