siRNA Details
Gene Symbol: NARF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038618.2 | NP_001033707.1 | 8, 9 | 1134 | ||
NM_001083608.1 | NP_001077077.1 | 7, 8 | 832 | |||
NM_012336.3 | NP_036468.1 | 8, 9 | 976 | |||
NM_031968.2 | NP_114174.1 | 9, 10 | 1114 | |||
XM_005256340.3 | XP_005256397.1 | 8, 9 | 1142 | |||
XM_006722278.3 | XP_006722341.1 | 7, 8 | 929 | |||
XM_011523541.2 | XP_011521843.1 | 8, 9 | 1066 | |||
XM_011523543.2 | XP_011521845.1 | 9, 10 | 979 | |||
XM_017024449.1 | XP_016879938.1 | 9, 10 | 1726 | |||
XM_017024450.1 | XP_016879939.1 | 9, 10 | 1216 | |||
GenBank mRNA | AF128406.1 | - | 8, 9 | 927 | ||
AK000929.1 | - | 8, 9 | 929 | |||
AK056966.1 | - | 8, 9 | 1109 | |||
BC000438.2 | - | 9, 10 | 1081 | 2820621 | ||
BC016440.1 | - | 7, 8 | 719 | 3447710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
