siRNA Details
Gene Symbol: CPEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177381.1 | NP_001170852.1 | 2 | 1770 | ||
NM_001177382.1 | NP_001170853.1 | 2 | 1770 | |||
NM_001177383.1 | NP_001170854.1 | 2 | 1770 | |||
NM_001177384.1 | NP_001170855.1 | 2 | 1770 | |||
NM_182485.2 | NP_872291.2 | 2 | 1770 | |||
NM_182646.2 | NP_872587.2 | 2 | 1770 | |||
XM_005248135.3 | XP_005248192.2 | 2 | 1993 | |||
XM_011513777.2 | XP_011512079.1 | 2 | 1993 | |||
XM_011513778.2 | XP_011512080.1 | 2 | 1993 | |||
XM_011513779.1 | XP_011512081.1 | 2 | 1993 | |||
XM_017007733.1 | XP_016863222.1 | 2 | 129 | |||
XM_017007734.1 | XP_016863223.1 | 2 | 130 | |||
GenBank mRNA | AY247744.1 | - | 2 | 546 | ||
AY255519.1 | - | 2 | 546 | |||
BC103939.1 | - | 3 | 442 | 40008488 | ||
BC103940.1 | - | 2 | 508 | 40008489 | ||
BC103941.1 | - | 2 | 511 | 40008490 | ||
BC103942.1 | - | 3 | 442 | 40008491 | ||
BC105925.1 | - | 2 | 508 | 40008495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
