siRNA Details
Gene Symbol: FERMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031471.5 | NP_113659.3 | 11 | 1399 | ||
NM_178443.2 | NP_848537.1 | 11 | 1411 | |||
XM_011545294.2 | XP_011543596.1 | 11 | 1423 | |||
XM_011545295.2 | XP_011543597.1 | 10 | 1256 | |||
XM_017018398.1 | XP_016873887.1 | 11 | 1407 | |||
XM_017018399.1 | XP_016873888.1 | 10 | 1259 | |||
GenBank mRNA | AK093719.1 | - | 11 | 1344 | ||
AY093951.1 | - | 11 | 1345 | |||
AY093952.1 | - | 11 | 1357 | |||
BC004347.1 | - | 11 | 1343 | 3634032 | ||
BC013366.2 | - | 11 | 1343 | 3677274 | ||
BC015584.2 | - | 11 | 1338 | 4637808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
