siRNA Details
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021950.3 | NP_068769.2 | 7 | 1618 | ||
NM_152866.2 | NP_690605.1 | 8 | 1881 | |||
GenBank mRNA | AK225630.1 | - | 7 | 1541 | ||
X07203.1 | - | 7 | 1556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
