siRNA Details
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021950.3 | NP_068769.2 | 7 | 1420 | ||
NM_152866.2 | NP_690605.1 | 8 | 1683 | |||
GenBank mRNA | AK225630.1 | - | 7 | 1343 | ||
AK300025.1 | - | 4 | 877 | |||
BC002807.2 | - | 7 | 1371 | 3634040 | ||
X07203.1 | - | 7 | 1357 | |||
X12530.1 | - | 7 | 1360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
