siRNA Details
Gene Symbol: ZNF274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278734.1 | NP_001265663.1 | 8 | 2152 | ||
NM_016324.3 | NP_057408.2 | 5 | 1836 | |||
NM_016325.3 | NP_057409.1 | 7 | 2041 | |||
NM_133502.2 | NP_598009.1 | 8 | 2137 | |||
XM_011526327.1 | XP_011524629.1 | 7 | 1575 | |||
XM_011526328.2 | XP_011524630.1 | 3 | 1256 | |||
XM_017026174.1 | XP_016881663.1 | 6 | 1842 | |||
XM_017026175.1 | XP_016881664.1 | 6 | 1997 | |||
GenBank mRNA | AB029149.1 | - | 5 | 1796 | ||
AB029150.1 | - | 7 | 1943 | |||
AF275680.1 | - | 8 | 1816 | |||
AF275681.1 | - | 2 | 1250 | |||
AK095162.1 | - | 2 | 2481 | |||
AK222855.1 | - | 5 | 1764 | |||
AK222878.1 | - | 5 | 1764 | |||
BC009763.2 | - | 7 | 1664 | 2966764 | ||
BT007304.1 | - | 7 | 1638 | |||
BX641169.1 | - | 2 | 4251 | |||
U71598.1 | - | 1 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
