siRNA Details
Gene Symbol: JAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002226.4 | NP_002217.3 | 25 | 3618 | ||
NM_145159.2 | NP_660142.1 | 24 | 3504 | |||
GenBank mRNA | AB209370.1 | - | 1 | 3636 | ||
AF003521.1 | - | 25 | 3246 | |||
AF020201.1 | - | 25 | 3265 | |||
AF029778.1 | - | 25 | 3618 | |||
AF029779.1 | - | 24 | 3504 | |||
LN607838.1 | - | 2 | 367 | |||
Y14330.1 | - | 25 | 3169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
