siRNA Details
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 5 | 975 | ||
NM_001098492.1 | NP_001091962.1 | 4 | 936 | |||
NM_001098493.1 | NP_001091963.1 | 4 | 933 | |||
NM_001098494.1 | NP_001091964.1 | 4 | 876 | |||
NM_001098495.1 | NP_001091965.1 | 3 | 837 | |||
NM_001098496.1 | NP_001091966.1 | 3 | 834 | |||
NM_001291743.1 | NP_001278672.1 | 4 | 873 | |||
NM_001291744.1 | NP_001278673.1 | 4 | 892 | |||
NM_024691.3 | NP_078967.3 | 5 | 972 | |||
GenBank mRNA | AB209371.1 | - | 3 | 4221 | ||
AK026886.1 | - | 4 | 846 | |||
AK302135.1 | - | 3 | 795 | |||
AK304569.1 | - | 4 | 881 | |||
AK307802.1 | - | 5 | 964 | |||
AK309427.1 | - | 3 | 1439 | |||
AK316156.1 | - | 4 | 894 | |||
AK316297.1 | - | 4 | 926 | |||
BC008297.1 | - | 5 | 918 | 3544044 | ||
BC024259.1 | - | 5 | 918 | 3604215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
