siRNA Details
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 5 | 2155 | ||
NM_001098492.1 | NP_001091962.1 | 4 | 2116 | |||
NM_001098493.1 | NP_001091963.1 | 4 | 2113 | |||
NM_001098494.1 | NP_001091964.1 | 4 | 2056 | |||
NM_001098495.1 | NP_001091965.1 | 3 | 2017 | |||
NM_001098496.1 | NP_001091966.1 | 3 | 2014 | |||
NM_001291743.1 | NP_001278672.1 | 4 | 2053 | |||
NM_001291744.1 | NP_001278673.1 | 4 | 2072 | |||
NM_001291745.1 | NP_001278674.1 | 4 | 604 | |||
NM_024691.3 | NP_078967.3 | 5 | 2152 | |||
GenBank mRNA | AB209371.1 | - | 3 | 5401 | ||
AK026886.1 | - | 4 | 2025 | |||
AK302135.1 | - | 3 | 1975 | |||
AK316156.1 | - | 4 | 2074 | |||
BC008297.1 | - | 5 | 2098 | 3544044 | ||
BC024259.1 | - | 5 | 2098 | 3604215 | ||
BF110540.1 | - | 4 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
