siRNA Details
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 5 | 2027 | ||
NM_001098492.1 | NP_001091962.1 | 4 | 1988 | |||
NM_001098493.1 | NP_001091963.1 | 4 | 1985 | |||
NM_001098494.1 | NP_001091964.1 | 4 | 1928 | |||
NM_001098495.1 | NP_001091965.1 | 3 | 1889 | |||
NM_001098496.1 | NP_001091966.1 | 3 | 1886 | |||
NM_001291743.1 | NP_001278672.1 | 4 | 1925 | |||
NM_001291744.1 | NP_001278673.1 | 4 | 1944 | |||
NM_024691.3 | NP_078967.3 | 5 | 2024 | |||
GenBank mRNA | AB209371.1 | - | 3 | 5273 | ||
AK026886.1 | - | 4 | 1897 | |||
AK302135.1 | - | 3 | 1847 | |||
AK304569.1 | - | 4 | 1933 | |||
AK316156.1 | - | 4 | 1946 | |||
BC008297.1 | - | 5 | 1970 | 3544044 | ||
BC024259.1 | - | 5 | 1970 | 3604215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
