siRNA Details
Gene Symbol: APOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136540.1 | NP_001130012.1 | 6 | 1239 | ||
NM_001136541.1 | NP_001130013.1 | 5 | 1146 | |||
NM_003661.3 | NP_003652.2 | 6 | 1200 | |||
NM_145343.2 | NP_663318.1 | 7 | 1354 | |||
XM_005261796.3 | XP_005261853.1 | 5 | 1125 | |||
XM_011530478.2 | XP_011528780.1 | 6 | 1272 | |||
GenBank mRNA | AF019225.2 | - | 6 | 1199 | ||
AF323540.1 | - | 7 | 1353 | |||
AK298053.1 | - | 3 | 930 | |||
AK300454.1 | - | 5 | 1115 | |||
AK303919.1 | - | 6 | 1173 | |||
AK309143.1 | - | 7 | 1385 | |||
AK313752.1 | - | 7 | 1293 | |||
BC017331.1 | - | 6 | 1130 | |||
BC112943.1 | - | 6 | 1155 | 4512933 | ||
BC113867.1 | - | 6 | 1130 | |||
BC114475.1 | - | 6 | 1130 | |||
BC127186.1 | - | 6 | 1130 | 40034503 | ||
BC141823.1 | - | 6 | 1155 | 40148156 | ||
BC142720.1 | - | 6 | 1155 | |||
BC143038.1 | - | 6 | 1130 | 40034505 | ||
BC143039.1 | - | 6 | 1130 | 40034506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
