siRNA Details
Gene Symbol: ZNF334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270497.1 | NP_001257426.1 | 6 | 522 | ||
NM_018102.4 | NP_060572.3 | 5 | 462 | |||
NR_073026.1 | 5 | 430 | ||||
XM_011528892.2 | XP_011527194.1 | 6 | 1967 | |||
XM_011528895.2 | XP_011527197.1 | 5 | 1457 | |||
XM_011528897.2 | XP_011527199.1 | 5 | 1762 | |||
XM_017027933.1 | XP_016883422.1 | 4 | 1575 | |||
XM_017027934.1 | XP_016883423.1 | 7 | 725 | |||
XM_017027935.1 | XP_016883424.1 | 6 | 1373 | |||
XM_017027936.1 | XP_016883425.1 | 5 | 1331 | |||
XM_017027937.1 | XP_016883426.1 | 4 | 1687 | |||
XM_017027938.1 | XP_016883427.1 | 5 | 375 | |||
XM_017027939.1 | XP_016883428.1 | 6 | 1475 | |||
XM_017027940.1 | XP_016883429.1 | 6 | 609 | |||
XM_017027941.1 | XP_016883430.1 | 5 | 1394 | |||
XM_017027942.1 | XP_016883431.1 | 4 | 1628 | |||
XM_017027943.1 | XP_016883432.1 | 4 | 1326 | |||
XM_017027944.1 | XP_016883433.1 | 3 | 1510 | |||
GenBank mRNA | AK001331.1 | - | 5 | 457 | ||
AK057717.1 | - | 4 | 752 | |||
AK095611.1 | - | 6 | 518 | |||
AL833936.1 | - | 4 | 639 | |||
BC024177.2 | - | 5 | 458 | 4824096 | ||
BC026321.1 | - | 6 | 1484 | 4819292 | ||
BG720863.1 | - | 5 | 463 | |||
DA080528.1 | - | 5 | 398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
