siRNA Details
Gene Symbol: LSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260489.1 | NP_001247418.1 | 2, 3 | 818 | ||
NM_015925.6 | NP_057009.3 | 2, 3 | 818 | |||
NM_205834.3 | NP_991403.1 | 2, 3 | 818 | |||
NM_205835.3 | NP_991404.1 | 2, 3 | 818 | |||
XM_005258980.1 | XP_005259037.1 | 2, 3 | 793 | |||
XM_005258982.1 | XP_005259039.1 | 2, 3 | 793 | |||
XM_011527026.1 | XP_011525328.1 | 2, 3 | 793 | |||
GenBank mRNA | AF130366.1 | - | 2, 3 | 484 | ||
AK075426.1 | - | 2, 3 | 470 | |||
AK126834.1 | - | 2, 3 | 1082 | |||
AK130367.1 | - | 2, 3 | 472 | |||
BC000015.2 | - | 2, 3 | 449 | |||
BC004381.1 | - | 2, 3 | 466 | |||
BC047376.1 | - | 2, 3 | 754 | 5272128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
