siRNA Details
Gene Symbol: LSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260489.1 | NP_001247418.1 | 1 | 237 | ||
NM_001260490.1 | NP_001247419.1 | 1 | 237 | |||
NM_015925.6 | NP_057009.3 | 1 | 237 | |||
NM_205834.3 | NP_991403.1 | 1 | 237 | |||
NM_205835.3 | NP_991404.1 | 1 | 237 | |||
XM_005258980.1 | XP_005259037.1 | 1 | 212 | |||
XM_005258982.1 | XP_005259039.1 | 1 | 212 | |||
XM_011527026.1 | XP_011525328.1 | 1 | 212 | |||
GenBank mRNA | AF130366.1 | - | 1 | 14 | ||
AK126834.1 | - | 1 | 501 | |||
BC047376.1 | - | 1 | 173 | 5272128 | ||
BM837160.1 | - | 1 | 139 | |||
BQ711340.1 | - | 1 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
