siRNA Details
Gene Symbol: CNTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256063.1 | NP_001242992.1 | 14 | 1863 | ||
NM_001256064.1 | NP_001242993.1 | 14 | 2076 | |||
NM_001843.3 | NP_001834.2 | 14 | 1863 | |||
NM_175038.2 | NP_778203.1 | 12 | 1679 | |||
XM_005268651.2 | XP_005268708.1 | 14 | 2226 | |||
XM_006719241.2 | XP_006719304.1 | 14 | 1850 | |||
XM_011537926.2 | XP_011536228.1 | 14 | 1815 | |||
XM_011537927.2 | XP_011536229.1 | 14 | 1872 | |||
XM_017018821.1 | XP_016874310.1 | 14 | 1866 | |||
XM_017018822.1 | XP_016874311.1 | 14 | 1815 | |||
XM_017018823.1 | XP_016874312.1 | 14 | 2226 | |||
XM_017018824.1 | XP_016874313.1 | 14 | 1850 | |||
XM_017018825.1 | XP_016874314.1 | 14 | 1815 | |||
XM_017018826.1 | XP_016874315.1 | 14 | 1850 | |||
XM_017018827.1 | XP_016874316.1 | 14 | 1872 | |||
GenBank mRNA | AK289544.1 | - | 14 | 1955 | ||
AK289698.1 | - | 14 | 1805 | |||
BC036569.2 | - | 14 | 1802 | 5273941 | ||
U07819.1 | - | 13 | 1636 | |||
U07820.1 | - | 12 | 1684 | |||
Z21488.1 | - | 14 | 1757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
