siRNA Details
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007226.1 | NP_001007227.1 | 6 | 723 | ||
NM_001007227.1 | NP_001007228.1 | 5 | 601 | |||
NM_001007228.1 | NP_001007229.1 | 4 | 549 | |||
NM_001007229.1 | NP_001007230.1 | 5 | 600 | |||
NM_001007230.1 | NP_001007231.1 | 6 | 652 | |||
NM_003563.3 | NP_003554.1 | 5 | 671 | |||
XM_005257723.4 | XP_005257780.1 | 5 | 895 | |||
XM_005257724.4 | XP_005257781.1 | 4 | 825 | |||
XM_017025204.1 | XP_016880693.1 | 3 | 338 | |||
GenBank mRNA | AJ000644.1 | - | 4 | 409 | ||
AK222589.1 | - | 4 | 374 | |||
AK293045.1 | - | 5 | 882 | |||
AK293587.1 | - | 4 | 491 | |||
AK300616.1 | - | 4 | 373 | |||
AK303268.1 | - | 3 | 295 | |||
AK312691.1 | - | 4 | 385 | |||
AK316366.1 | - | 6 | 450 | |||
BC001269.1 | - | 5 | 555 | 3461565 | ||
BC003385.1 | - | 4 | 366 | 3445878 | ||
CB992730.1 | - | 6 | 624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
