siRNA Details
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007226.1 | NP_001007227.1 | 6 | 700 | ||
NM_001007227.1 | NP_001007228.1 | 5 | 578 | |||
NM_001007228.1 | NP_001007229.1 | 4 | 526 | |||
NM_001007229.1 | NP_001007230.1 | 5 | 577 | |||
NM_001007230.1 | NP_001007231.1 | 6 | 629 | |||
NM_003563.3 | NP_003554.1 | 5 | 648 | |||
XM_005257723.4 | XP_005257780.1 | 5 | 872 | |||
XM_005257724.4 | XP_005257781.1 | 4 | 802 | |||
XM_017025204.1 | XP_016880693.1 | 3 | 315 | |||
GenBank mRNA | AJ000644.1 | - | 4 | 386 | ||
AK222589.1 | - | 4 | 351 | |||
AK293587.1 | - | 4 | 468 | |||
AK300616.1 | - | 4 | 350 | |||
AK303268.1 | - | 3 | 272 | |||
AK312691.1 | - | 4 | 362 | |||
AK316366.1 | - | 6 | 427 | |||
BC001269.1 | - | 5 | 532 | 3461565 | ||
BC003385.1 | - | 4 | 343 | 3445878 | ||
CB992730.1 | - | 6 | 601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
