siRNA Details
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007226.1 | NP_001007227.1 | 10 | 1282 | ||
NM_001007227.1 | NP_001007228.1 | 9 | 1160 | |||
NM_001007228.1 | NP_001007229.1 | 8 | 1108 | |||
NM_001007229.1 | NP_001007230.1 | 9 | 1159 | |||
NM_001007230.1 | NP_001007231.1 | 10 | 1211 | |||
NM_003563.3 | NP_003554.1 | 9 | 1230 | |||
XM_005257723.4 | XP_005257780.1 | 9 | 1454 | |||
XM_005257724.4 | XP_005257781.1 | 8 | 1384 | |||
XM_017025204.1 | XP_016880693.1 | 7 | 897 | |||
GenBank mRNA | AJ000644.1 | - | 8 | 968 | ||
AK222589.1 | - | 8 | 933 | |||
AK293045.1 | - | 9 | 1441 | |||
AK293587.1 | - | 8 | 914 | |||
AK300616.1 | - | 8 | 932 | |||
AK303268.1 | - | 7 | 854 | |||
AK312691.1 | - | 8 | 944 | |||
AK316366.1 | - | 10 | 1009 | |||
BC001269.1 | - | 9 | 1114 | 3461565 | ||
BC003385.1 | - | 8 | 925 | 3445878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
