siRNA Details
Gene Symbol: MYOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013451.3 | NP_038479.1 | 7 | 816 | ||
NM_133337.2 | NP_579899.1 | 7 | 816 | |||
XM_005269693.4 | XP_005269750.1 | 7 | 995 | |||
XM_005269694.4 | XP_005269751.1 | 7 | 995 | |||
XM_011539632.2 | XP_011537934.1 | 8 | 1189 | |||
XM_017016068.1 | XP_016871557.1 | 6 | 576 | |||
XM_017016069.1 | XP_016871558.1 | 6 | 621 | |||
XM_017016070.1 | XP_016871559.1 | 7 | 1003 | |||
GenBank mRNA | AB033033.2 | - | 7 | 693 | ||
AF182316.1 | - | 7 | 789 | |||
AF207990.1 | - | 7 | 693 | |||
BC040110.1 | - | 8 | 1006 | |||
BC052617.1 | - | 7 | 781 | 6422729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
