siRNA Details
Gene Symbol: UBE3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014671.2 | NP_055486.2 | 19 | 2941 | ||
XM_005249564.4 | XP_005249621.1 | 18 | 2875 | |||
XM_017012818.1 | XP_016868307.1 | 20 | 3213 | |||
GenBank mRNA | AK127280.1 | - | 4 | 3212 | ||
AK295138.1 | - | 19 | 2621 | |||
D13635.1 | - | 19 | 2932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
