siRNA Details
Gene Symbol: UBE3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014671.2 | NP_055486.2 | 3 | 483 | ||
XM_017012818.1 | XP_016868307.1 | 4 | 755 | |||
GenBank mRNA | AK295138.1 | - | 3 | 163 | ||
BC014029.2 | - | 4 | 243 | 4541634 | ||
D13635.1 | - | 3 | 474 | |||
DB454915.1 | - | 3 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
