siRNA Details
Gene Symbol: ARID4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002892.3 | NP_002883.3 | 16 | 1679 | ||
NM_023000.2 | NP_075376.2 | 16 | 1679 | |||
NM_023001.2 | NP_075377.2 | 16 | 1679 | |||
XM_005267964.2 | XP_005268021.1 | 16 | 1798 | |||
XM_005267965.2 | XP_005268022.1 | 16 | 1798 | |||
XM_017021560.1 | XP_016877049.1 | 16 | 1798 | |||
XM_017021561.1 | XP_016877050.1 | 16 | 1575 | |||
XM_017021562.1 | XP_016877051.1 | 16 | 1798 | |||
XM_017021563.1 | XP_016877052.1 | 16 | 1798 | |||
XM_017021564.1 | XP_016877053.1 | 14 | 1406 | |||
XM_017021565.1 | XP_016877054.1 | 6 | 1841 | |||
GenBank mRNA | AK307520.1 | - | 16 | 1628 | ||
AK307972.1 | - | 658 | ||||
S57153.1 | - | 4 | 414 | |||
S57160.1 | - | 4 | 414 | |||
S57162.1 | - | 4 | 414 | |||
S66427.1 | - | 16 | 1539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
