siRNA Details
Gene Symbol: C21orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004649.7 | NP_004640.4 | 7 | 836 | ||
NM_198155.4 | NP_937798.4 | 6 | 743 | |||
NR_135220.1 | 6 | 783 | ||||
XM_017028479.1 | XP_016883968.1 | 6 | 647 | |||
GenBank mRNA | BC002370.2 | - | 7 | 821 | 2822340 | |
BC003587.1 | - | 7 | 767 | 2822714 | ||
BQ882034.1 | - | 6 | 640 | |||
CX788692.1 | - | 6 | 735 | |||
D86061.1 | - | 7 | 767 | |||
D86062.1 | - | 6 | 675 | |||
U53003.1 | - | 7 | 834 | |||
Y07572.1 | - | 7 | 817 |
Gene Symbol: LOC102724023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723901.3 | XP_006723964.1 | 7 | 859 | ||
XM_006723903.3 | XP_006723966.1 | 6 | 753 | |||
XM_011546081.2 | XP_011544383.1 | 6 | 730 | |||
XM_017028517.1 | XP_016884006.1 | 6 | 647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
