siRNA Details
Gene Symbol: ZPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198844.2 | NP_942141.2 | 4 | 626 | ||
NM_199321.2 | NP_955353.1 | 5 | 692 | |||
XM_011524298.1 | XP_011522600.1 | 5 | 730 | |||
GenBank mRNA | AK292383.1 | - | 5 | 730 | ||
AY251603.1 | - | 5 | 675 | |||
AY251604.1 | - | 4 | 609 | |||
BC043152.2 | - | 4 | 612 | 5297840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
