siRNA Details
Gene Symbol: PRNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000311.3 | NP_000302.1 | 2 | 1260 | ||
NM_001080121.1 | NP_001073590.1 | 2 | 1255 | |||
NM_001080122.1 | NP_001073591.1 | 2 | 1251 | |||
NM_001080123.1 | NP_001073592.1 | 2 | 1124 | |||
NM_001271561.1 | NP_001258490.1 | 2 | 1260 | |||
NM_183079.2 | NP_898902.1 | 2 | 1256 | |||
GenBank mRNA | AB300823.1 | - | 1 | 898 | ||
AB300824.1 | - | 1 | 898 | |||
AK090575.1 | - | 3 | 859 | |||
AK293312.1 | - | 3 | 858 | |||
AK295415.1 | - | 3 | 810 | |||
AK295943.1 | - | 3 | 876 | |||
AY008282.1 | - | 2 | 931 | |||
BC012844.1 | - | 2 | 935 | 3863784 | ||
BC016809.1 | - | 1 | 88 | |||
BC022532.1 | - | 2 | 948 | 4813982 | ||
D00015.1 | - | 1 | 940 | |||
M13667.1 | - | 1 | 940 | |||
M13899.1 | - | 2 | 937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
