siRNA Details
Gene Symbol: PRNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000311.3 | NP_000302.1 | 2 | 679 | ||
NM_001080121.1 | NP_001073590.1 | 2 | 674 | |||
NM_001080122.1 | NP_001073591.1 | 2 | 670 | |||
NM_001080123.1 | NP_001073592.1 | 2 | 543 | |||
NM_001271561.1 | NP_001258490.1 | 2 | 679 | |||
NM_183079.2 | NP_898902.1 | 2 | 675 | |||
GenBank mRNA | AB300823.1 | - | 1 | 317 | ||
AB300824.1 | - | 1 | 317 | |||
AB300825.1 | - | 1 | 317 | |||
AB300826.1 | - | 1 | 317 | |||
AK293312.1 | - | 3 | 277 | |||
AK295415.1 | - | 3 | 229 | |||
AK295943.1 | - | 3 | 295 | |||
AK312276.1 | - | 2 | 367 | |||
AY008282.1 | - | 2 | 350 | |||
AY569456.1 | - | 1 | 307 | |||
BC012844.1 | - | 2 | 354 | 3863784 | ||
BC022532.1 | - | 2 | 367 | 4813982 | ||
BG397054.1 | - | 407 | ||||
BI669189.1 | - | 2 | 370 | |||
BP251427.1 | - | 2 | 456 | |||
BT019496.1 | - | 1 | 307 | |||
CR542039.1 | - | 1 | 307 | |||
CR542072.1 | - | 1 | 307 | |||
D00015.1 | - | 1 | 362 | |||
M13667.1 | - | 1 | 362 | |||
M13899.1 | - | 2 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
