siRNA Details
Gene Symbol: TBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005996.3 | NP_005987.3 | 3 | 1697 | ||
NM_016569.3 | NP_057653.3 | 4 | 1757 | |||
GenBank mRNA | AF002228.1 | - | 3 | 733 | ||
AF170708.2 | - | 3 | 993 | |||
AF216750.1 | - | 4 | 909 | |||
AK054604.1 | - | 3 | 1700 | |||
AY034105.1 | - | 2 | 179 | |||
BC025258.1 | - | 4 | 1183 | 5458526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
