siRNA Details
Gene Symbol: PUF60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136033.2 | NP_001129505.1 | 12 | 1897 | ||
NM_001271096.1 | NP_001258025.1 | 11 | 1824 | |||
NM_001271097.1 | NP_001258026.1 | 10 | 1740 | |||
NM_001271098.1 | NP_001258027.1 | 12 | 1875 | |||
NM_001271099.1 | NP_001258028.1 | 11 | 1791 | |||
NM_001271100.1 | NP_001258029.1 | 11 | 1846 | |||
NM_014281.4 | NP_055096.2 | 11 | 1827 | |||
NM_078480.2 | NP_510965.1 | 12 | 1878 | |||
XM_011516929.2 | XP_011515231.1 | 13 | 2077 | |||
XM_011516930.2 | XP_011515232.1 | 12 | 1928 | |||
XM_017013234.1 | XP_016868723.1 | 12 | 3410 | |||
XM_017013235.1 | XP_016868724.1 | 12 | 1980 | |||
XM_017013236.1 | XP_016868725.1 | 11 | 3359 | |||
XM_017013237.1 | XP_016868726.1 | 13 | 2139 | |||
XM_017013238.1 | XP_016868727.1 | 13 | 1939 | |||
XM_017013239.1 | XP_016868728.1 | 12 | 1905 | |||
XM_017013240.1 | XP_016868729.1 | 11 | 1854 | |||
GenBank mRNA | AF114818.1 | - | 12 | 1859 | ||
AF190744.1 | - | 12 | 1787 | |||
AF217197.2 | - | 11 | 1754 | |||
BC008875.2 | - | 11 | 1793 | 4138778 | ||
BC009734.1 | - | 11 | 1776 | 3877563 | ||
BC011265.1 | - | 12 | 1814 | 4155549 | ||
BC011979.1 | - | 10 | 1693 | 3863660 | ||
BC035435.2 | - | 11 | 1778 | |||
U51586.1 | - | 10 | 1741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
