siRNA Details
Gene Symbol: HPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007216.3 | NP_009147.3 | 16 | 2643 | ||
NM_181507.1 | NP_852608.1 | 17 | 2800 | |||
NM_181508.1 | NP_852609.1 | 16 | 2456 | |||
XM_011519862.1 | XP_011518164.1 | 17 | 2791 | |||
XM_011519863.1 | XP_011518165.1 | 17 | 2699 | |||
XM_011519864.1 | XP_011518166.1 | 17 | 2631 | |||
XM_011519865.1 | XP_011518167.1 | 16 | 3013 | |||
XM_011519866.1 | XP_011518168.1 | 16 | 2664 | |||
XM_011519867.1 | XP_011518169.1 | 16 | 2530 | |||
XM_011519868.2 | XP_011518170.1 | 16 | 2771 | |||
XM_017017149.1 | XP_016872638.1 | 17 | 2699 | |||
XM_017017150.1 | XP_016872639.1 | 17 | 2631 | |||
XM_017017151.1 | XP_016872640.1 | 16 | 2849 | |||
XM_017017152.1 | XP_016872641.1 | 16 | 3013 | |||
XM_017017153.1 | XP_016872642.1 | 16 | 2849 | |||
XM_017017154.1 | XP_016872643.1 | 16 | 2530 | |||
GenBank mRNA | AB023234.1 | - | 16 | 2456 | ||
AF534400.1 | - | 16 | 2456 | |||
AF534401.1 | - | 17 | 2800 | |||
AF534402.1 | - | 16 | 2643 | |||
AJ131721.1 | - | 5 | 974 | |||
AK024320.1 | - | 2 | 583 | |||
AK291663.1 | - | 17 | 2766 | |||
AK292436.1 | - | 16 | 2609 | |||
BC033640.1 | - | 16 | 2604 | 5502950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
