siRNA Details
Gene Symbol: HPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007216.3 | NP_009147.3 | 22 | 3534 | ||
NM_181507.1 | NP_852608.1 | 23 | 3691 | |||
NM_181508.1 | NP_852609.1 | 22 | 3347 | |||
XM_011519862.1 | XP_011518164.1 | 24 | 3816 | |||
XM_011519863.1 | XP_011518165.1 | 24 | 3724 | |||
XM_011519864.1 | XP_011518166.1 | 24 | 3656 | |||
XM_011519865.1 | XP_011518167.1 | 23 | 4038 | |||
XM_011519866.1 | XP_011518168.1 | 23 | 3689 | |||
XM_011519867.1 | XP_011518169.1 | 23 | 3555 | |||
XM_011519868.2 | XP_011518170.1 | 23 | 3796 | |||
XM_017017149.1 | XP_016872638.1 | 23 | 3590 | |||
XM_017017150.1 | XP_016872639.1 | 23 | 3522 | |||
XM_017017151.1 | XP_016872640.1 | 23 | 3874 | |||
XM_017017152.1 | XP_016872641.1 | 22 | 3904 | |||
XM_017017153.1 | XP_016872642.1 | 22 | 3740 | |||
XM_017017154.1 | XP_016872643.1 | 22 | 3421 | |||
GenBank mRNA | AB023234.1 | - | 22 | 3347 | ||
AF534400.1 | - | 22 | 3347 | |||
AF534401.1 | - | 23 | 3691 | |||
AF534402.1 | - | 22 | 3534 | |||
AJ131721.1 | - | 11 | 1865 | |||
AK024320.1 | - | 8 | 1474 | |||
AK292436.1 | - | 22 | 3500 | |||
BC033640.1 | - | 22 | 3495 | 5502950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
