siRNA Details
Gene Symbol: RREB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003698.3 | NP_001003698.1 | 10 | 4039 | ||
NM_001003699.3 | NP_001003699.1 | 10 | 4039 | |||
NM_001003700.1 | NP_001003700.1 | 10 | 4039 | |||
NM_001168344.1 | NP_001161816.1 | 10 | 4003 | |||
XM_006715157.3 | XP_006715220.1 | 11 | 10876 | |||
XM_011514794.2 | XP_011513096.1 | 9 | 3903 | |||
XM_011514795.2 | XP_011513097.1 | 10 | 4116 | |||
GenBank mRNA | AB019254.1 | - | 10 | 3862 | ||
AB019351.1 | - | 10 | 3862 | |||
AB019352.1 | - | 10 | 3862 | |||
AF072825.1 | - | 1 | 67 | |||
AF072826.1 | - | 1 | 67 | |||
D49835.1 | - | 11 | 4002 | |||
HM369361.1 | - | 7 | 3547 | |||
U26914.1 | - | 1 | 1013 |

More Information
Other Species Targeted

Gene Ontology Categories:
