siRNA Details
Gene Symbol: RREB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003698.3 | NP_001003698.1 | 10 | 4094 | ||
NM_001003699.3 | NP_001003699.1 | 10 | 4094 | |||
NM_001003700.1 | NP_001003700.1 | 10 | 4094 | |||
NM_001168344.1 | NP_001161816.1 | 10 | 4058 | |||
XM_006715157.3 | XP_006715220.1 | 11 | 10931 | |||
XM_011514794.2 | XP_011513096.1 | 9 | 3958 | |||
XM_011514795.2 | XP_011513097.1 | 10 | 4171 | |||
GenBank mRNA | AB019254.1 | - | 10 | 3917 | ||
AB019351.1 | - | 10 | 3917 | |||
AB019352.1 | - | 10 | 3917 | |||
AF072825.1 | - | 1 | 122 | |||
AF072826.1 | - | 1 | 122 | |||
D49835.1 | - | 11 | 4057 | |||
HM369361.1 | - | 7 | 3602 | |||
U26914.1 | - | 1 | 1068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
