siRNA Details
Gene Symbol: CALB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001740.4 | NP_001731.2 | 2, 3 | 258 | ||
NM_007088.3 | NP_009019.1 | 2, 3 | 258 | |||
NR_027910.2 | 2, 3 | 258 | ||||
GenBank mRNA | AK222495.1 | - | 2, 3 | 248 | ||
AK222648.1 | - | 2, 3 | 248 | |||
AK291096.1 | - | 2, 3 | 248 | |||
BC015484.2 | - | 2, 3 | 233 | 3847342 | ||
DA060805.1 | - | 2, 3 | 258 | |||
X56667.1 | - | 2, 3 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
